N-2,3-Dihydro-1H-inden-5-ylacetamide ≥97%

Supplier: Matrix Scientific

Synonyms: N-(2,3-Dihydro-1H-inden-5-yl)acetamide

081470-1G 081470-10G
102835-012EA 200 USD
102835-012 102835-014
N-2,3-Dihydro-1H-inden-5-ylacetamide ≥97%
N-2,3-Dihydro-1H-inden-5-ylacetamide

Hazard: Irritant

Formula: C₁₁H₁₃NO
MW: 175.23 g/mol
MDL Number: MFCD00221464
CAS Number: 59856-06-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR