(±)-3-Aminoquinuclidine dihydrochloride ≥98%
Supplier: Matrix Scientific
Synonyms:
3-Aminoquinuclidine dihydrochloride, 3-Amino-1-azabicyclo[2.2.2]octane dihydrochloride
081439-1G
081439-10G
081439-50G
102834-938EA
101.54
USD
102834-938
102834-940
102834-942
(±)-3-Aminoquinuclidine dihydrochloride ≥98%
(±)-3-Aminoquinuclidine dihydrochloride
Hazard: Irritant
Formula:
C₇H₁₆Cl₂N₂ MW: 199.1213 g/mol Melting Pt: >280 °C |
MDL Number:
MFCD00137395 CAS Number: 6530-09-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...