(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: (R)-3-Aminoquinuclidine dihydrochloride

081437-1G 081437-10G
102834-928EA 46.15 USD
102834-928 102834-930
(R)-(+)-3-Aminoquinuclidine dihydrochloride ≥97%
(R)-(+)-3-Aminoquinuclidine dihydrochloride

Hazard: Irritant

Formula: C₇H₁₆Cl₂N₂
MW: 199.1213 g/mol
MDL Number: MFCD00191752
CAS Number: 123536-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR