5-(Methylthio)-1,2,4-thiadiazol-3-amine ≥97%

Supplier: Matrix Scientific

081435-1G 081435-10G
102834-920EA 138.46 USD
102834-920 102834-922
5-(Methylthio)-1,2,4-thiadiazol-3-amine ≥97%
5-(Methylthio)-1,2,4-thiadiazol-3-amine

Hazard: Irritant

Formula: C₃H₅N₃S₂
MDL Number: MFCD00799288
CAS Number: 60093-10-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR