3-Amino-1,2,4-triazine ≥97%

Supplier: Matrix Scientific

081433-1G 081433-10G
102834-914EA 101.54 USD
102834-914 102834-916
3-Amino-1,2,4-triazine ≥97%
3-Amino-1,2,4-triazine

Hazard: Irritant

Formula: C₃H₄N₄
MDL Number: MFCD00006459
CAS Number: 1120-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR