3-Amino-5-methylthio-1H-1,2,4-triazole ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-5-(methylmercapto)-1H-1,2,4-triazole

081424-1G 081424-10G
102834-882EA 101.54 USD
102834-882 102834-884
3-Amino-5-methylthio-1H-1,2,4-triazole ≥97%
3-Amino-5-methylthio-1H-1,2,4-triazole

Hazard: Irritant

Formula: C₃H₆N₄S
MDL Number: MFCD00005232
CAS Number: 45534-08-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR