2-Amino-5-ethyl-1,3,4-thiadiazole ≥97%
Supplier: Matrix Scientific
Synonyms:
5-Ethyl-1,3,4-thiadiazol-2-amine
081422-1G
081422-10G
081422-50G
102834-874EA
56.92
USD
102834-874
102834-876
102834-878
2-Amino-5-ethyl-1,3,4-thiadiazole ≥97%
2-Amino-5-ethyl-1,3,4-thiadiazole
Hazard: Irritant
Formula:
C₄H₇N₃S MW: 129.19 g/mol |
MDL Number:
MFCD00003111 CAS Number: 14068-53-2 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...