5-(Methylthio)-1,3,4-thiadiazol-2-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-(methylthio)-1,3,4-thiadiazole

081421-1G 081421-10G
102834-870EA 143.08 USD
102834-870 102834-872
5-(Methylthio)-1,3,4-thiadiazol-2-amine ≥97%
5-(Methylthio)-1,3,4-thiadiazol-2-amine

Hazard: Irritant

Formula: C₃H₅N₃S₂
MDL Number: MFCD00175752
CAS Number: 5319-77-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR