2-Amino-5-methyl-1,3,4-thiadiazole ≥97%

Supplier: Matrix Scientific

081420-1G 081420-10G
102834-866EA 56.92 USD
102834-866 102834-868
2-Amino-5-methyl-1,3,4-thiadiazole ≥97%
2-Amino-5-methyl-1,3,4-thiadiazole

Hazard: Irritant

Formula: C₃H₅N₃S
MW: 115.16 g/mol
MDL Number: MFCD00003110
CAS Number: 108-33-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR