tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥97%

Supplier: Matrix Scientific

081419-1G 081419-10G
102834-862EA 298.46 USD
102834-862 102834-864
tert-Butyl methyl(pyrrolidin-3-yl)carbamate ≥97%
tert-Butyl methyl(pyrrolidin-3-yl)carbamate

Hazard: Irritant

Formula: C₁₀H₂₀N₂O₂
MDL Number: MFCD00191665
CAS Number: 172478-00-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR