3-Aminopyrrolidine dihydrochloride ≥97%

Supplier: Matrix Scientific

Synonyms: Pyrrolidin-3-amine dihydrochloride

081418-1G 081418-10G
102834-858EA 101.54 USD
102834-858 102834-860
3-Aminopyrrolidine dihydrochloride ≥97%
3-Aminopyrrolidine dihydrochloride

Hazard: Irritant

Formula: C₄H₁₀N₂
MW: 159.05 g/mol
MDL Number: MFCD00060176
CAS Number: 103831-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR