1-Benzyl-3-aminopyrrolidine ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Amino-1-benzylpyrrolidine, 1-Benzylpyrrolidin-3-amine

081417-1G 081417-10G
102834-854EA 44.62 USD
102834-854 102834-856
1-Benzyl-3-aminopyrrolidine ≥97%
1-Benzyl-3-aminopyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00059036
CAS Number: 18471-40-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR