(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥97%

Supplier: Matrix Scientific

Synonyms: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

081414-1G 081414-10G
102834-846EA 101.54 USD
102834-846 102834-848
(3R)-(-)-1-Benzyl-3-aminopyrrolidine ≥97%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine

Hazard: Irritant

Formula: C₁₁H₁₆N₂
MW: 176.26 g/mol
MDL Number: MFCD00082638
CAS Number: 114715-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR