5-(2-Furyl)-1,3,4-oxadiazol-2-amine ≥97%

Supplier: Matrix Scientific

081412-1G 081412-10G
102834-840EA 255.38 USD
102834-840 102834-842
5-(2-Furyl)-1,3,4-oxadiazol-2-amine ≥97%
5-(2-Furyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₆H₅N₃O₂
MW: 151.13 g/mol
Melting Pt: 225…226 °C
MDL Number: MFCD01710630
CAS Number: 7659-06-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR