5-(4-Nitrophenyl)-1,3,4-oxadiazol-2-amine ≥97%

Supplier: Matrix Scientific

081410-1G 081410-10G
102834-834EA 283.08 USD
102834-834 102834-836
5-(4-Nitrophenyl)-1,3,4-oxadiazol-2-amine ≥97%
5-(4-Nitrophenyl)-1,3,4-oxadiazol-2-amine

Hazard: Irritant

Formula: C₈H₆N₄O₃
MW: 206.16 g/mol
MDL Number: MFCD00603997
CAS Number: 51891-79-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR