3-Aminopiperidine ≥97%
Supplier: Matrix Scientific
Synonyms:
Piperidin-3-amine
081404-1G
081404-10G
102834-818EA
83.08
USD
102834-818
102834-820
3-Aminopiperidine ≥97%
3-Aminopiperidine
Hazard: Corrosive/Stored under Argon
Formula:
C₅H₁₂N₂ MW: 100.16 g/mol Boiling Pt: 170°C Melting Pt: 57 °C |
MDL Number:
MFCD04972497 CAS Number: 54012-73-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...