4-Aminopiperidine ≥97%

Supplier: Matrix Scientific

Synonyms: Piperidin-4-amine

081403-1G 081403-10G
102834-814EA 156.92 USD
102834-814 102834-816
4-Aminopiperidine ≥97%
4-Aminopiperidine

Hazard: Irritant

Formula: C₅H₁₂N₂
MW: 100.16 g/mol
MDL Number: MFCD02179399
CAS Number: 13035-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR