1-Benzylpiperidin-4-amine dihydrochloride ≥97%

Supplier: Matrix Scientific

081400-1G 081400-10G 081400-50G
102834-804EA 86.15 USD
102834-804 102834-806 102834-808
1-Benzylpiperidin-4-amine dihydrochloride ≥97%
1-Benzylpiperidin-4-amine dihydrochloride

Hazard: Irritant

Formula: C₁₂H₂₀Cl₂N₂
MDL Number: MFCD00044825
CAS Number: 1205-72-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR