6-Nitroquinolin-5-ylamine ≥97%

Supplier: Matrix Scientific

081390-1G 081390-10G
102834-782EA 152.31 USD
102834-782 102834-784
6-Nitroquinolin-5-ylamine ≥97%
6-Nitroquinolin-5-ylamine

Hazard: Irritant

Formula: C₉H₇N₃O₂
MDL Number: MFCD00006798
CAS Number: 35975-00-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR