2-Aminobenzimidazole ≥97%
Supplier: Matrix Scientific
081386-10G
081386-50G
102834-772EA
116.92
USD
102834-772
102834-774
2-Aminobenzimidazole ≥97%
2-Aminobenzimidazole
Hazard: Irritant
Formula:
C₇H₇N₃ |
MDL Number:
MFCD00005596 CAS Number: 934-32-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...