2-Amino-5-chlorobenzoxazole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chlorobenzo[d]oxazol-2-amine, Zoxazolamine, 2-Amino-5-chloro-1,3-benzoxazole

081385-1G 081385-10G
102834-768EA 72.31 USD
102834-768 102834-770
2-Amino-5-chlorobenzoxazole ≥97%
2-Amino-5-chlorobenzoxazole

Hazard: Irritant

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD00005770
CAS Number: 61-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR