5-Chloro-2-methyl-1,3-benzoxazol-6-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-chloro-6-benzoxazolamine

081383-1G 081383-10G
102834-762EA 327.69 USD
102834-762 102834-764
5-Chloro-2-methyl-1,3-benzoxazol-6-amine ≥97%
5-Chloro-2-methyl-1,3-benzoxazol-6-amine

Hazard: Irritant

Formula: C₈H₇ClN₂O
MW: 182.61 g/mol
MDL Number: MFCD07368636
CAS Number: 323579-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR