1-[(4-Methylphenyl)sulfonyl]-1H-indol-5-amine hydrochloride ≥85%

Supplier: Matrix Scientific

Synonyms: 5-Amino-1-[(4-methylphenyl)sulphonyl]-1H-indole hydrochloride

081380-1G 081380-10G
102834-754EA 553.85 USD
102834-754 102834-756
1-[(4-Methylphenyl)sulfonyl]-1H-indol-5-amine hydrochloride ≥85%
1-[(4-Methylphenyl)sulfonyl]-1H-indol-5-amine hydrochloride

Hazard: Irritant

Formula: C₁₅H₁₅ClN₂O₂S
MW: 322.82 g/mol
MDL Number: MFCD02259420
CAS Number: 306937-24-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR