6-Amino-1,3-dimethyluracil ≥85%

Supplier: Matrix Scientific

Synonyms: 6-Amino-1,3-dimethylpyrimidine-2,4(1H,3H)-dione, 4-Amino-1,3-Dimethyluracil

081375-1G 081375-10G 081375-50G
102834-738EA 56.92 USD
102834-738 102834-740 102834-742
6-Amino-1,3-dimethyluracil ≥85%
6-Amino-1,3-dimethyluracil

Hazard: Irritant

Formula: C₆H₉N₃O₂
MW: 155.1546 g/mol
MDL Number: MFCD00006552
CAS Number: 6642-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR