4-Amino-2-bromopyrimidine-5-carbonitrile ≥97%

Supplier: Matrix Scientific

081359-1G 081359-10G
102834-698EA 124.62 USD
102834-698 102834-700
4-Amino-2-bromopyrimidine-5-carbonitrile ≥97%
4-Amino-2-bromopyrimidine-5-carbonitrile

Hazard: Irritant

Formula: C₅H₃BrN₄
MW: 199.01 g/mol
MDL Number: MFCD00173661
CAS Number: 94741-70-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR