2-Chloro-N-methylpyrimidin-4-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(methylamino)pyrimidine

081354-1G 081354-10G
102834-686EA 229.23 USD
102834-686 102834-688
2-Chloro-N-methylpyrimidin-4-amine ≥97%
2-Chloro-N-methylpyrimidin-4-amine

Hazard: Irritant

Formula: C₅H₆ClN₃
MW: 143.58 g/mol
MDL Number: MFCD09055368
CAS Number: 66131-68-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR