3-Amino-4-bromo-5-phenyl-1H-pyrazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-5-phenyl-1H-pyrazol-3-amine, (3-Amino-4-bromo-1H-pyrazol-5-yl)benzene

081348-1G 081348-10G
102834-672EA 127.69 USD
102834-672 102834-674
3-Amino-4-bromo-5-phenyl-1H-pyrazole ≥97%
3-Amino-4-bromo-5-phenyl-1H-pyrazole

Hazard: Irritant

Formula: C₉H₈BrN₃
MW: 238.09 g/mol
MDL Number: MFCD00082666
CAS Number: 2845-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR