3-Amino-1-phenyl-2-pyrazolin-5-one ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Amino-2,4-dihydro-2-phenyl-3H-pyrazol-3-one

081345-10G 081345-50G
102834-664EA 64.62 USD
102834-664 102834-666
3-Amino-1-phenyl-2-pyrazolin-5-one ≥97%
3-Amino-1-phenyl-2-pyrazolin-5-one

Hazard: Irritant

Formula: C₉H₉N₃O
MW: 175.19 g/mol
Melting Pt: 218…219 °C
MDL Number: MFCD00003137
CAS Number: 4149-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR