1-Methyl-3-thien-2-yl-1H-pyrazol-5-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 1-Methyl-3-(thiophen-2-yl)-1H-pyrazol-5-amine, 5-Amino-1-methyl-3-(2-thienyl)pyrazole

081344-1G 081344-10G
102834-660EA 184.62 USD
102834-660 102834-662
1-Methyl-3-thien-2-yl-1H-pyrazol-5-amine ≥97%
1-Methyl-3-thien-2-yl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₈H₉N₃S
MW: 179.25 g/mol
MDL Number: MFCD00068075
CAS Number: 118430-78-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR