3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine ≥97%

Supplier: Matrix Scientific

081342-1G 081342-10G
102834-652EA 127.69 USD
102834-652 102834-654
3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine ≥97%
3-Cyclopropyl-1-methyl-1H-pyrazol-5-amine

Hazard: Irritant

Formula: C₇H₁₁N₃
MDL Number: MFCD00067984
CAS Number: 118430-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR