3-Amino-5-methylisoxazole ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Methylisoxazol-3-amine, 3-Amino-5-methylisoxazol, 5-Methylisoxazol-3-ylamine

081330-1G 081330-10G
102834-624EA 56.92 USD
102834-624 102834-626
3-Amino-5-methylisoxazole ≥97%
3-Amino-5-methylisoxazole

Hazard: Irritant

Formula: C₄H₆N₂O
MW: 98.1 g/mol
Boiling Pt: 194
MDL Number: MFCD00003155
CAS Number: 1072-67-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR