5-Amino-3,4-dimethylisoxazole ≥97%

Supplier: Matrix Scientific

081327-1G 081327-10G
102834-616EA 56.92 USD
102834-616 102834-618
5-Amino-3,4-dimethylisoxazole ≥97%
5-Amino-3,4-dimethylisoxazole

Hazard: Irritant

Formula: C₅H₈N₂O
MDL Number: MFCD00046081
CAS Number: 19947-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR