4-(4-Trifluoromethylphenyl)thiazol-2-ylamine ≥97%

Supplier: Matrix Scientific

081325-1G 081325-10G
102834-610EA 200 USD
102834-610 102834-612
4-(4-Trifluoromethylphenyl)thiazol-2-ylamine ≥97%
4-(4-Trifluoromethylphenyl)thiazol-2-ylamine

Hazard: Irritant

Formula: C₁₀H₇F₃N₂S
MW: 244.24 g/mol
MDL Number: MFCD03094298
CAS Number: 105512-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR