2-Amino-4-methylthiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Methylthiazol-2-ylamine, 4-Methylthiazol-2-amine, 2-Amino-4-methylthiazol

081323-1G 081323-10G 081323-50G
102834-602EA 56.92 USD
102834-602 102834-604 102834-606
2-Amino-4-methylthiazole ≥97%
2-Amino-4-methylthiazole

Hazard: Irritant

Formula: C₄H₆N₂S
MW: 114.17 g/mol
MDL Number: MFCD00005329
CAS Number: 1603-91-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR