3-Amino-2-phenylpyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Phenylpyridin-3-amine
081288-1G
081288-10G
102834-526EA
383.08
USD
102834-526
102834-528
3-Amino-2-phenylpyridine ≥97%
3-Amino-2-phenylpyridine
Hazard: Irritant
Formula:
C₁₁H₁₀N₂ MW: 170.22 g/mol |
MDL Number:
MFCD04114113 CAS Number: 101601-80-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...