6-Aminonicotinamide ≥97%
Supplier: Matrix Scientific
Synonyms:
6-Aminopyridine-3-carboxamide
081287-1G
081287-10G
081287-50G
102834-520EA
113.85
USD
102834-520
102834-522
102834-524
6-Aminonicotinamide ≥97%
6-Aminonicotinamide
Hazard: Irritant
Formula:
C₆H₇N₃O MW: 137.14 g/mol |
MDL Number:
MFCD00006327 CAS Number: 329-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...