2,6-Dimethoxypyridin-3-amine hydrochloride ≥97%

Supplier: Matrix Scientific

081276-1G 081276-10G
102834-496EA 298.46 USD
102834-496 102834-498
2,6-Dimethoxypyridin-3-amine hydrochloride ≥97%
2,6-Dimethoxypyridin-3-amine hydrochloride

Hazard: Irritant

Formula: C₇H₁₁ClN₂O₂
MDL Number: MFCD00012816
CAS Number: 80789-72-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR