3-Amino-2-(4-fluorophenoxy)pyridine ≥97%
Supplier: Matrix Scientific
081274-1G
081274-10G
102834-490EA
149.23
USD
102834-490
102834-492
3-Amino-2-(4-fluorophenoxy)pyridine ≥97%
3-Amino-2-(4-fluorophenoxy)pyridine
Hazard: Irritant
Formula:
C₁₁H₉FN₂O |
MDL Number:
MFCD00067803 CAS Number: 175135-64-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...