6-Phenoxypyridin-3-amine ≥97%

Supplier: Matrix Scientific

081273-1G 081273-10G
102834-486EA 241.54 USD
102834-486 102834-488
6-Phenoxypyridin-3-amine ≥97%
6-Phenoxypyridin-3-amine

Hazard: Irritant

Formula: C₁₁H₁₀N₂O
MW: 186.22 g/mol
MDL Number: MFCD01692449
CAS Number: 25194-67-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR