2-(Boc-amino)-5-bromopyridine ≥97%
Supplier: Matrix Scientific
081257-1G
081257-10G
102834-452EA
72.31
USD
102834-452
102834-454
2-(Boc-amino)-5-bromopyridine ≥97%
2-(Boc-amino)-5-bromopyridine
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂O₂ |
MDL Number:
MFCD06411381 CAS Number: 159451-66-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...