5-(Boc-amino)-2-bromopyridine ≥97%
Supplier: Matrix Scientific
Synonyms:
tert-Butyl (6-bromopyridin-3-yl)carbamate, 5-Amino-2-bromopyridine, 5-BOC protected
081255-1G
081255-10G
102834-446EA
298.46
USD
102834-446
102834-448
5-(Boc-amino)-2-bromopyridine ≥97%
5-(Boc-amino)-2-bromopyridine
Hazard: Irritant
Formula:
C₁₀H₁₃BrN₂O₂ MW: 273.13 g/mol |
MDL Number:
MFCD07363808 CAS Number: 218594-15-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...