4-(Boc-amino)-2-chloropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Chloro-4-(tert-butoxycarbonyl)pyridine, tert-Butyl (2-chloropyridin-4-yl)carbamate, 4-Amino-2-chloropyridine, 4-BOC protected, tert-Butyl 2-chloropyridine-4-carbamate

081250-1G 081250-10G
102834-434EA 213.85 USD
102834-434 102834-436
4-(Boc-amino)-2-chloropyridine ≥97%
4-(Boc-amino)-2-chloropyridine

Hazard: Irritant

Formula: C₁₀H₁₃ClN₂O₂
MW: 228.68
MDL Number: MFCD02179232
CAS Number: 234108-73-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR