3-Amino-2-bromo-4,6-dimethylpyridine ≥97%
Supplier: Matrix Scientific
081247-1G
081247-10G
102834-426EA
383.08
USD
102834-426
102834-428
3-Amino-2-bromo-4,6-dimethylpyridine ≥97%
3-Amino-2-bromo-4,6-dimethylpyridine
Hazard: Irritant
Formula:
C₇H₉BrN₂ |
MDL Number:
MFCD03731181 CAS Number: 104829-98-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...