2-Methoxy-6-(methylamino)pyridine ≥97%

Supplier: Matrix Scientific

081246-1G 081246-10G
102834-422EA 72.31 USD
102834-422 102834-424
2-Methoxy-6-(methylamino)pyridine ≥97%
2-Methoxy-6-(methylamino)pyridine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MDL Number: MFCD00191557
CAS Number: 88569-83-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR