2-Aminopyridine-N-oxide ≥97%
Supplier: Matrix Scientific
Synonyms:
2-Aminopyridine 1-oxide, 2-Aminopyridine N-Oxide
081223-1G
081223-10G
102834-372EA
549.23
USD
102834-372
102834-374
2-Aminopyridine-N-oxide ≥97%
2-Aminopyridine N-oxide
Hazard: Irritant
Formula:
C₅H₆N₂O |
MDL Number:
MFCD00059168 CAS Number: 14150-95-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...