2-Amino-5-methyl-3-nitropyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Amino-5-nitro-3-picoline, 2-Amino-3-nitro-5-picoline, 5-Methyl-3-nitro-2-pyridylamine, 5-Methyl-3-nitropyridin-2-amine

081222-1G 081222-10G
102834-368EA 184.62 USD
102834-368 102834-370
2-Amino-5-methyl-3-nitropyridine ≥97%
2-Amino-5-methyl-3-nitropyridine

Hazard: Irritant

Formula: C₆H₇N₃O₂
MW: 153.14 g/mol
MDL Number: MFCD00047442
CAS Number: 7598-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR