2-Ethoxypyridin-4-amine ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Amino-2-ethoxypyridine

081215-1G 081215-10G
102834-352EA 467.69 USD
102834-352 102834-354
2-Ethoxypyridin-4-amine ≥97%
2-Ethoxypyridin-4-amine

Hazard: Irritant

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
MDL Number: MFCD00235127
CAS Number: 89943-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR