3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide ≥95%
Supplier: Matrix Scientific
081211-1G
081211-10G
102834-344EA
383.08
USD
102834-344
102834-346
3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide ≥95%
3-Oxo-N-(3-(trifluoromethyl)phenyl)butanamide
Hazard: Irritant
Formula:
C₁₁H₁₀F₃NO₂ MW: 245.2 g/mol |
MDL Number:
MFCD00201876 CAS Number: 785-74-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...