2-Bromo-4-fluoro-6-nitroaniline ≥97%
Supplier: Matrix Scientific
081208-1G
081208-5G
102834-336EA
116.92
USD
102834-336
102834-338
2-Bromo-4-fluoro-6-nitroaniline ≥97%
2-Bromo-4-fluoro-6-nitroaniline
Hazard: Toxic
Formula:
C₆H₄BrFN₂O₂ MW: 235.01 g/mol Melting Pt: 74-75°C |
MDL Number:
MFCD07779528 CAS Number: 10472-88-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...