3,4-Dichloro-N-methylaniline ≥95%
Supplier: Matrix Scientific
Synonyms:
N1-Methyl-3,4-dichloroaniline
081191-1G
081191-10G
102834-298EA
86.15
USD
102834-298
102834-300
3,4-Dichloro-N-methylaniline ≥95%
3,4-Dichloro-N-methylaniline
Hazard: Irritant
Formula:
C₇H₇Cl₂N MW: 176.04 g/mol |
MDL Number:
MFCD00052850 CAS Number: 40750-59-2 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...